1,3,5-Tri(9H-carbazol-9-yl)benzene ≥98.0% (by HPLC), purified by sublimation
Leverandør: TCI
T1934-1G
T1934-5G
TCIAT1934-1GEA
1925
NOK
TCIAT1934-1G
TCIAT1934-5G
1,3,5-Tri(9H-carbazol-9-yl)benzene ≥98.0% (by HPLC), purified by sublimation
1,3,5-Tri(9H-carbazol-9-yl)benzene
Formel:
C₄₂H₂₇N₃ MW: 573,70 g/mol |
MDL nummer:
MFCD03844810 CAS-nummer: 148044-07-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...