1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
N,N'-(Methylenebis(4,1-phenylene))bis(2,2-dimethylhydrazinecarboxamide), Bis[4-[3-(dimethylamino)ureido]phenyl]methane
T2051-25G
T2051-500G
TCIAT2051-25GEA
350
NOK
TCIAT2051-25G
TCIAT2051-500G
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide ≥95.0% (by HPLC, titration analysis)
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide
Formel:
C₁₉H₂₆N₆O₂ MW: 370,46 g/mol |
MDL nummer:
MFCD00525631 CAS-nummer: 85095-61-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 174.0 to 179.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...