2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2-N,2-N,4-N,4-N,6-N,6-N-hexakis(methoxymethyl)-1,3,5-triazine-2,4,6-triamine, Hexakis(methoxymethyl)melamine, N,N,N',N',N'',N''-Hexakis(methoxymethyl)melamine

T2059-25G T2059-5G
TCIAT2059-25GEA 2700 NOK
TCIAT2059-25G TCIAT2059-5G
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine ≥98.0% (by GC)
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine
Formel: C₁₅H₃₀N₆O₆
MW: 390,44 g/mol
MDL nummer: MFCD00047342
CAS-nummer: 3089-11-0
EINECS: 221-422-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3adba4fec21ae5f2410d37083f2f04d10a8f7923&vid=3adba4fec21ae5f2410d37083f2f04d10a8f7923&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 53.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR