2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: (3R,4S,5S,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-galactose

T2112-1G T2112-5G
TCIAT2112-1GEA 1325 NOK
TCIAT2112-1G TCIAT2112-5G
2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98.0% (by HPLC)
2,3,4,6-Tetra-O-benzyl-D-galactopyranose
Formel: C₃₄H₃₆O₆
MW: 540,65 g/mol
MDL nummer: MFCD00132927
CAS-nummer: 6386-24-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of isomer)
Melting point 63.0 to 67.0 deg-C
Specific rotation [a]20/D +9.0 to +13.0 deg (C=1,CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR