Trimethoprim ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

T2286-25G T2286-100G T2286-500G
TCIAT2286-25GEA 1175 NOK
TCIAT2286-25G TCIAT2286-100G TCIAT2286-500G
Trimethoprim ≥98.0% (by HPLC, titration analysis)
Trimethoprim
Formel: C₁₄H₁₈N₄O₃
MW: 290,32 g/mol
MDL nummer: MFCD00036761
CAS-nummer: 738-70-5
EINECS: 212-006-2
Merck Indeks: 14,09709
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e35965d50481a261179e2454e53e0807cc3d018d&vid=e35965d50481a261179e2454e53e0807cc3d018d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 199.0 to 205.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR