4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide ≥95.0% (by GC)

Leverandør: TCI
Warning

T2645-1G T2645-5G
TCIAT2645-1GEA 1025 NOK
TCIAT2645-1G TCIAT2645-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide ≥95.0% (by GC)
4,4,5,5-Tetramethyl-1,3,2-dioxaphospholane-2-oxide
Formel: C₆H₁₃O₃P
MW: 164,14 g/mol
MDL nummer: MFCD09836231
CAS-nummer: 16352-18-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 102.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR