2,2,2-Trichloroethyl sulphamate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Sulphamic acid 2,2,2-Trichloroethyl Ester, 2,2,2-Trichloroethoxysulphonamide

T2646-5G T2646-1G
TCIAT2646-5GEA 2200 NOK
TCIAT2646-5G TCIAT2646-1G
2,2,2-Trichloroethyl sulphamate ≥98.0% (by titrimetric analysis)
2,2,2-Trichloroethyl sulphamate
Formel: C₂H₄Cl₃NO₃S
MW: 228,47 g/mol
MDL nummer: MFCD01683146
CAS-nummer: 69226-51-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR