1,3,5-Tris(4-carboxyphenyl)benzene ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 5'-(4-Carboxyphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

T2647-1G T2647-5G
TCIAT2647-1GEA 1400 NOK
TCIAT2647-1G TCIAT2647-5G
1,3,5-Tris(4-carboxyphenyl)benzene ≥98.0% (by HPLC)
1,3,5-Tris(4-carboxyphenyl)benzene
Formel: C₂₇H₁₈O₆
MW: 438,44 g/mol
Smeltepunkt: 325°C
MDL nummer: MFCD10000888
CAS-nummer: 50446-44-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR