2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Leverandør: TCI
T2700-1G
T2700-5G
TCIAT2700-1GEA
2500
NOK
TCIAT2700-1G
TCIAT2700-5G
2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine
Formel:
C₃₉H₂₄N₆ MW: 576,66 g/mol |
MDL nummer:
MFCD18207722 CAS-nummer: 134984-37-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...