Tropisetron hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (3-endo)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 1H-Indole-3-carboxylate Hydrochloride, ICS 25-93, SDZ ICS-93, 3-Tropanylindole-3-carboxylate hydrochloride, endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-yl 1H-indole-3-carboxylate hydrochloride

T2743-5G T2743-1G
TCIAT2743-5GEA 3125 NOK
TCIAT2743-5G TCIAT2743-1G
Tropisetron hydrochloride ≥98.0% (by HPLC, titration analysis)
Tropisetron hydrochloride
Formel: C₁₇H₂₀N₂O₂
MW: 320,82 g/mol
Smeltepunkt: 285°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00210221
CAS-nummer: 105826-92-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,09783
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6c70511554e42db26e3f6bdc4767b0aef445e1a&vid=e6c70511554e42db26e3f6bdc4767b0aef445e1a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR