5-Thiazolemethanol ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Thiazol-5-ylmethanol, 5-(Hydroxymethyl)thiazole, Thiazole-5-methanol

T2779-25G T2779-5G
TCIAT2779-25GEA 2000 NOK
TCIAT2779-25G TCIAT2779-5G
5-Thiazolemethanol ≥98.0% (by GC)
5-Thiazolemethanol
Formel: C₄H₅NOS
MW: 115,15 g/mol
Kokepunkt: 106°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD04115732
CAS-nummer: 38585-74-9
EINECS: 414-780-9

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR