2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
|
Danger
|
T2806-1G
T2806-100MG
TCIAT2806-1GEA
4925
NOK
TCIAT2806-1G
TCIAT2806-100MG
2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene ≥98.0% (by HPLC, total nitrogen)
2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene
Formel:
C₁₈H₁₆N₄S₈ MW: 544,83 g/mol |
CAS-nummer:
132765-36-7 FN: 3439 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Orange to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...