2-Thiophenesulphonamide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Thiophene-2-sulphonamide

T2851-1G T2851-5G
TCIAT2851-1GEA 405 NOK
TCIAT2851-1G TCIAT2851-5G
2-Thiophenesulphonamide ≥98.0% (by GC)
2-Thiophenesulphonamide
Formel: C₄H₅NO₂S₂
MW: 163,21 g/mol
MDL nummer: MFCD00185853
CAS-nummer: 6339-87-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR