3-Trimethylsilylpropynal ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(Trimethylsilyl)-2-propynal
T2907-1G
T2907-5G
TCIAT2907-1GEA
1700
NOK
TCIAT2907-1G
TCIAT2907-5G
3-Trimethylsilylpropynal ≥97.0% (by GC)
3-Trimethylsilylpropynal
Formel:
C₆H₁₀OSi MW: 126,23 g/mol Kokepunkt: 45°C Flash punkt: 29°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD01103709 CAS-nummer: 2975-46-4 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...