3-(Trifluoromethyl)benzenesulphonamide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
α,α,α-Trifluoro-m-toluenesulphonamide
T2925-1G
T2925-5G
TCIAT2925-1GEA
365
NOK
TCIAT2925-1G
TCIAT2925-5G
3-(Trifluoromethyl)benzenesulphonamide ≥98.0% (by HPLC, titration analysis)
3-(Trifluoromethyl)benzenesulphonamide
Formel:
C₇H₆F₃NO₂S MW: 225,19 g/mol |
MDL nummer:
MFCD00042421 CAS-nummer: 672-58-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...