Tyrphostin AG 490 98,0 HPLC, T

Leverandør: TCI
Danger

Synonymer: AG 490, ag 490, AG-490, tyrphostin ag 490, 2-Cyano-3-(3,4-dihydroxyphenyl)-N-(phenylmethyl)-2-propenamide, (E)-N-Benzyl-2-cyano-3-(3,4-dihydroxyphenyl)acrylamide, Tyrphostin AG 490, AG490, 2-Cyano-3-(3,4-dihydroxyphenyl)-N-(benzyl)-2-propenamide

T2962-20MG T2962-100MG T2962-1G
TCIAT2962-20MGEA 460 NOK
TCIAT2962-20MG TCIAT2962-100MG TCIAT2962-1G
Tyrphostin AG 490 98,0 HPLC, T
Tyrphostin AG 490
Formel: C₁₇H₁₄N₂O₃
MW: 294.31 g/mol
Smeltepunkt: 215°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00236452
CAS-nummer: 133550-30-8
FN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c10a11afc49a48571ac8640cfa4cd118f9799325&vid=c10a11afc49a48571ac8640cfa4cd118f9799325&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR