4-(Trimethylsilyl)-3-butyn-2-one ≥97.0% (by GC)

Leverandør: TCI
Warning

T2973-5ML T2973-25ML
TCIAT2973-5MLEA 785 NOK
TCIAT2973-5ML TCIAT2973-25ML
4-(Trimethylsilyl)-3-butyn-2-one ≥97.0% (by GC)
4-(Trimethylsilyl)-3-butyn-2-one
Formel: C₇H₁₂OSi
Kokepunkt: 158°C
Flash punkt: 28°C
MDL nummer: MFCD00190212
CAS-nummer: 5930-98-3
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=903da6e7330b09b0fc80ef0fa665b596b587368a&vid=903da6e7330b09b0fc80ef0fa665b596b587368a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR