1-(2-(Trifluoromethyl)phenyl)imidazole ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(2-(Trifluoromethyl)phenyl)-1H-imidazole, TRIM
T2982-1G
T2982-200MG
TCIAT2982-1GEA
2850
NOK
TCIAT2982-1G
TCIAT2982-200MG
1-(2-(Trifluoromethyl)phenyl)imidazole ≥98.0% (by GC)
1-(2-(Trifluoromethyl)phenyl)imidazole
Formel:
C₁₀H₇F₃N₂ MW: 212,17 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00041206 CAS-nummer: 25371-96-4 |
Spesifikasjon Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...