Trimethyl(pentafluorophenyl)silane ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Trimethyl(perfluorophenyl)silane

T3012-1G T3012-5G
TCIAT3012-1GEA 380 NOK
TCIAT3012-1G TCIAT3012-5G
Trimethyl(pentafluorophenyl)silane ≥98.0% (by GC)
Trimethyl(pentafluorophenyl)silane
Formel: C₉H₉F₅Si
MW: 240,25 g/mol
Kokepunkt: 165°C
MDL nummer: MFCD00092630
CAS-nummer: 1206-46-8
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c658a858b5c2c8211fe564c1c1966cf5d42dea09&vid=c658a858b5c2c8211fe564c1c1966cf5d42dea09&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR