3-Aminophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

T3028-1G T3028-5G
TCIAT3028-1GEA 245 NOK
TCIAT3028-1G TCIAT3028-5G
3-Aminophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Aminophenylboronic acid pinacol ester
Formel: C₁₂H₁₈BNO₂
MW: 219,09 g/mol
MDL nummer: MFCD03453668
CAS-nummer: 210907-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7998fa6a118911e48c56e069d723c4cdc09444a&vid=a7998fa6a118911e48c56e069d723c4cdc09444a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR