4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (4'-Hydroxybiphenyl-4-yl)boronic Acid Pinacol Ester, 2-(4'-Hydroxybiphenyl-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3031-1G T3031-5G
TCIAT3031-1GEA 1275 NOK
TCIAT3031-1G TCIAT3031-5G
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol ≥98.0% (by GC)
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)biphenyl-4-ol
Formel: C₁₈H₂₁BO₃
MW: 296,17 g/mol
MDL nummer: MFCD22414472
CAS-nummer: 760989-91-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01dd377d91b021adb46c578c8a37ea50004d8348&vid=01dd377d91b021adb46c578c8a37ea50004d8348&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %
Melting point 147.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR