3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 3,3',5,5'-Tetra-tert-butylbiphenyl-2,2'-diol, 3,3',5,5'-Tetra-tert-butyl-[1,1'-biphenyl]-2,2'-diol

T3052-1G T3052-5G
TCIAT3052-1GEA 620 NOK
TCIAT3052-1G TCIAT3052-5G
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl ≥98.0% (by GC)
3,3',5,5'-Tetra-tert-butyl-2,2'-dihydroxybiphenyl
Formel: C₂₈H₄₂O₂
MW: 410,64 g/mol
MDL nummer: MFCD00085287
CAS-nummer: 6390-69-8
EINECS: 407-920-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a317434e0e284dec1c365cfa8ff31ff4eaaea86d&vid=a317434e0e284dec1c365cfa8ff31ff4eaaea86d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR