Tolnaftate ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: O-2-Naphthyl Methyl(3-methylphenyl)carbamothioate, O-Naphthalen-2-yl methyl(m-tolyl)carbamothioate, Methyl(3-methylphenyl)carbamothioic Acid O-2-Naphthyl Ester

T3059-1G T3059-5G
TCIAT3059-1GEA 350 NOK
TCIAT3059-1G TCIAT3059-5G
Tolnaftate ≥97.0% (by HPLC, total nitrogen)
Tolnaftate
Formel: C₁₉H₁₇NOS
MW: 307,41 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056611
CAS-nummer: 2398-96-1
EINECS: 219-266-6
Merck Indeks: 14,09519
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1b5ea911f41566f0ecccdbb3ce486793225ece7&vid=e1b5ea911f41566f0ecccdbb3ce486793225ece7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 111.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR