Triethyl orthobenzoate ≥97.0% (by GC)

Leverandør: TCI

Synonymer: alpha,alpha,alpha-Triethoxytoluene, (Triethoxymethyl)benzene, Orthobenzoic acid triethyl ester

T3070-5G T3070-25G
TCIAT3070-5GEA 350 NOK
TCIAT3070-5G TCIAT3070-25G
Triethyl orthobenzoate ≥97.0% (by GC)
Triethyl orthobenzoate
Formel: C₁₃H₂₀O₃
MW: 224,3 g/mol
Kokepunkt: 240°C
Flash punkt: 97°C
MDL nummer: MFCD00009222
CAS-nummer: 1663-61-2
EINECS: 216-771-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a950ff1de8ce9412a8250852cc143db334d929c&vid=1a950ff1de8ce9412a8250852cc143db334d929c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR