Ticlopidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: Triclopidine hydrochloride, Ticlopidine HCl, 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride

T3110-5G T3110-25G
TCIAT3110-5GEA 525 NOK
TCIAT3110-5G TCIAT3110-25G
Ticlopidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ticlopidine hydrochloride
Formel: C₁₄H₁₄ClNS
MW: 300.24 g/mol
MDL nummer: MFCD00079606
CAS-nummer: 53885-35-1
EINECS: 258-837-4
Merck Indeks: 14,09421
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02d2e82711b5d3ca3f27648933f83881f4e921b9&vid=02d2e82711b5d3ca3f27648933f83881f4e921b9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 208.0 to 212.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR