4-(Trifluoromethyl)benzamidine hydrochloride ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 4-(Trifluoromethyl)benzimidamide hydrochloride

T3115-200MG T3115-1G
TCIAT3115-200MGEA 1100 NOK
TCIAT3115-200MG TCIAT3115-1G
4-(Trifluoromethyl)benzamidine hydrochloride ≥98.0% (by HPLC)
4-(Trifluoromethyl)benzamidine hydrochloride
Formel: C₈H₇F₃N₂·HCl
MW: 224.61 g/mol
Smeltepunkt: 167°C
MDL nummer: MFCD04114431
CAS-nummer: 38980-96-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d422efc182901b0deca0d60c4803a116d4c5aac&vid=5d422efc182901b0deca0d60c4803a116d4c5aac&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR