ɑ-(Trichloromethyl)benzyl acetate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2,2,2-Trichloro-1-phenylethyl acetate, Acetic acid 2,2,2-Trichloro-1-phenylethyl Ester, Acetic Acid ɑ-(Trichloromethyl)benzyl Ester, Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester, 2,2,2-Trichloro-1-phenylethyl Acetate, Acetic acid α-(Trichloromethyl)benzyl Ester, α-(Trichloromethyl)benzyl Acetate

T3122-25G T3122-250G
TCIAT3122-25GEA 220 NOK
TCIAT3122-25G TCIAT3122-250G
ɑ-(Trichloromethyl)benzyl acetate ≥98.0% (by GC)
alpha-(Trichloromethyl)benzyl acetate
Formel: C₁₀H₉Cl₃O₂
MW: 267,53 g/mol
Kokepunkt: 282°C
MDL nummer: MFCD00000804
CAS-nummer: 90-17-5
EINECS: 201-972-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c831d24dfa21dc39a51183d9faa519050ce5c32d&vid=c831d24dfa21dc39a51183d9faa519050ce5c32d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR