2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl ≥98.0% (by GC, titration analysis) free radical
Leverandør: TCI
|
Warning
|
Synonymer:
4-Propargyloxy-TEMPO, 4-(2-Propynyloxy)-TEMPO, 2,2,6,6-Tetramethyl-4-(propargyloxy)piperidine 1-Oxyl
T3169-1G
T3169-5G
TCIAT3169-1GEA
1050
NOK
TCIAT3169-1G
TCIAT3169-5G
2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl ≥98.0% (by GC, titration analysis) free radical
2,2,6,6-Tetramethyl-4-(2-propynyloxy)piperidine 1-oxyl
Formel:
C₁₂H₂₀NO₂ MW: 210,30 g/mol |
MDL nummer:
MFCD01126982 CAS-nummer: 147045-24-7 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...