Tris(2-benzimidazolylmethyl)amine ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (BimH)3, Tris((1H-benzo[d]imidazol-2-yl)methyl)amine

T3170-200MG T3170-1G
TCIAT3170-200MGEA 620 NOK
TCIAT3170-200MG TCIAT3170-1G
Tris(2-benzimidazolylmethyl)amine ≥96.0% (by HPLC, titration analysis)
Tris(2-benzimidazolylmethyl)amine
Formel: C₂₄H₂₁N₇
MW: 407,48 g/mol
MDL nummer: MFCD00075516
CAS-nummer: 64019-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5abb61bce9639e580588e22adf9203bd506e9ef2&vid=5abb61bce9639e580588e22adf9203bd506e9ef2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR