Tiaramide hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
5-Chloro-3-[[4-(2-hydroxyethyl)piperazin-1-yl]carbonylmethyl]-2-benzothiazolinone hydrochloride, 5-Chloro-3-(2-(4-(2-hydroxyethyl)piperazin-1-yl)-2-oxoethyl)benzo[d]thiazol-2(3H)-one hydrochloride
T3176-25MG
T3176-100MG
TCIAT3176-25MGEA
1800
NOK
TCIAT3176-25MG
TCIAT3176-100MG
Tiaramide hydrochloride ≥98.0% (by titrimetric analysis)
Tiaramide hydrochloride
Formel:
C₁₅H₁₈ClN₃O₃S·HCl MW: 392,30 g/mol Smeltepunkt: 160°C |
MDL nummer:
MFCD01702832 CAS-nummer: 35941-71-0 EINECS: 252-802-7 Merck Indeks: 14,09424 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...