Tris(2,2,2-trifluoroethyl) Phosphate ≥96.0% (by GC)
Leverandør: TCI
Warning
Synonymer: Tris(2,2,2-trifluoroethyl)phosphate, Phosphoric acid tris(2,2,2-trifluoroethyl) ester
T3203-5G T3203-25G
TCIAT3203-5GEA 960 NOK
TCIAT3203-5G TCIAT3203-25G
Tris(2,2,2-trifluoroethyl) Phosphate ≥96.0% (by GC)
Tris(2,2,2-trifluoroethyl) Phosphate
Formel: C₆H₆F₉O₄P
MW: 344,07 g/mol
Kokepunkt: 40°C
Smeltepunkt: –22°C
MDL nummer: MFCD00233117
CAS-nummer: 358-63-4
FN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6513c8fe2c2d6a72c5b32c3fa4d4cab4c25e159&vid=b6513c8fe2c2d6a72c5b32c3fa4d4cab4c25e159&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %