N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer:
N2,N4,N6-tri-m-tolyl-1,3,5-Triazine-2,4,6-triamine, Tris(3-methylanilino)triazine
T3240-200MG
T3240-1G
TCIAT3240-200MGEA
485
NOK
TCIAT3240-200MG
TCIAT3240-1G
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine ≥98.0% (by HPLC, total nitrogen)
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine
Formel:
C₂₄H₂₄N₆ MW: 396,50 g/mol Smeltepunkt: 233°C |
MDL nummer:
MFCD19440763 CAS-nummer: 82504-70-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...