N,N-Dimethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,2,2-Trifluoro-N,N-dimethylacetamide

T3262-25G T3262-5G
TCIAT3262-25GEA 1875 NOK
TCIAT3262-25G TCIAT3262-5G
N,N-Dimethyl-2,2,2-trifluoroacetamide ≥98.0% (by GC)
N,N-Dimethyl-2,2,2-trifluoroacetamide
Formel: C₄H₆F₃NO
MW: 141,0900 g/mol
Kokepunkt: 136°C
MDL nummer: MFCD00043555
CAS-nummer: 1547-87-1
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR