1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
TMS-EBX
T3272-1G
T3272-5G
TCIAT3272-1GEA
1725
NOK
TCIAT3272-1G
TCIAT3272-5G
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by titrimetric analysis)
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one
Formel:
C₁₂H₁₃IO₂Si MW: 344,22 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD29765750 CAS-nummer: 181934-29-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Potassium iodate Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...