1,4,5,8-Tetrachloroanthraquinone ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,4,5,8-Tetrachloroanthracene-9,10-dione

T3300-5G T3300-25G
TCIAT3300-5GEA 1350 NOK
TCIAT3300-5G TCIAT3300-25G
1,4,5,8-Tetrachloroanthraquinone ≥95.0% (by GC)
1,4,5,8-Tetrachloroanthraquinone
Formel: C₁₄H₄Cl₄O₂
MW: 345,98 g/mol
Smeltepunkt: 342°C
MDL nummer: MFCD00035801
CAS-nummer: 81-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14561fb3de6043b7c1c5f33f70c6f2240ca9498&vid=b14561fb3de6043b7c1c5f33f70c6f2240ca9498&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR