Tetrandine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
(4aS,16aS)-3,4,4a,5,16a,17,18,19-octahydro-12,21,22,26-tetramethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2',3':17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinoline, (1beta)-6,6',7,12-Tetramethoxy-2,2'-dimethylberbaman, (11S,31S)-16,36,37,54-Tetramethoxy-12,32-dimethyl-11,12,13,14,31,32,33,34-octahydro-2,6-dioxa-1(7,1),3(8,1)-diisoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, Tetrandrine, NSC 77037, d-Tetrandine
T3321-200MG
T3321-1G
TCIAT3321-200MGEA
770
NOK
TCIAT3321-200MG
TCIAT3321-1G
Tetrandine ≥98.0% (by HPLC, titration analysis)
Tetrandine
Formel:
C₃₈H₄₂N₂O₆ MW: 622,8 g/mol |
MDL nummer:
MFCD08689909 CAS-nummer: 518-34-3 Merck Indeks: 14,09231 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 221.0 to 225.0 deg-C |
Specific rotation [a]20/D | +283 to +287 deg (C=1,CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...