1,2-Bis(diethylamino)ethane ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Danger
Synonymer: N1,N1,N2,N2-Tetraethylethane-1,2-diamine, N,N,N',N'-Tetraethylethylenediamine
T3371-5ML T3371-25ML
TCIAT3371-5MLEA 460 NOK
TCIAT3371-5ML TCIAT3371-25ML
1,2-Bis(diethylamino)ethane ≥98.0% (by GC, titration analysis)
1,2-Bis(diethylamino)ethane
Formel: C₁₀H₂₄N₂
MW: 172,31 g/mol
Kokepunkt: 190°C
Flash punkt: 58°C
MDL nummer: MFCD00009055
CAS-nummer: 150-77-6
EINECS: 205-770-3
FN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=543c6f45f41261b41d61060a9544bbd625b69b03&vid=543c6f45f41261b41d61060a9544bbd625b69b03&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %