2-Phenyl-2',4',6'-trihydroxyacetophenone ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2',4',6'-Trihydroxy-2-phenylacetophenone, 2-Phenyl-1-(2,4,6-trihydroxyphenyl)ethanone, Benzyl 2,4,6-Trihydroxyphenyl ketone
T3390-1G
T3390-5G
TCIAT3390-1GEA
875
NOK
TCIAT3390-1G
TCIAT3390-5G
2-Phenyl-2',4',6'-trihydroxyacetophenone ≥96.0% (by GC)
2-Phenyl-2',4',6'-trihydroxyacetophenone
Formel:
C₁₄H₁₂O₄ MW: 244,2427 g/mol |
MDL nummer:
MFCD00205539 CAS-nummer: 727-71-9 FN: 3077 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...