1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
T3439-200MG
T3439-1G
TCIAT3439-200MGEA
960
NOK
TCIAT3439-200MG
TCIAT3439-1G
1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene ≥98.0% (by HPLC, total nitrogen)
1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene
Formel:
C₆₀H₃₉N₃ MW: 801,99 g/mol |
MDL nummer:
MFCD29765751 CAS-nummer: 160780-82-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...