[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Mesityl(3-(trifluoromethyl)phenyl)iodonium trifluoromethanesulphonate, Mesityl[3-(trifluoromethyl)phenyl]iodonium triflate

T3445-1G T3445-5G
TCIAT3445-1GEA 1350 NOK
TCIAT3445-1G TCIAT3445-5G
[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
[3-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulphonate
Formel: C₁₇H₁₅F₆IO₃S
MW: 540,26 g/mol
Smeltepunkt: 183°C
MDL nummer: MFCD20264880
CAS-nummer: 1204518-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee384ac87093d6fb874e5d449415755c9a9ae800&vid=ee384ac87093d6fb874e5d449415755c9a9ae800&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR