trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 3,4,5-Trifluoro-4'-[(trans,trans)-4'-pentyl[1,1'-bicyclohexyl]-4-yl]-1,1'-biphenyl

T3482-1G T3482-5G
TCIAT3482-1GEA 500 NOK
TCIAT3482-1G TCIAT3482-5G
trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl
Formel: C₂₉H₃₇F₃
MW: 442,61 g/mol
MDL nummer: MFCD11053475
CAS-nummer: 137529-43-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR