Tetrahexylammonium hydrogen sulphate ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Warning

T3495-5G T3495-25G
TCIAT3495-5GEA 1225 NOK
TCIAT3495-5G TCIAT3495-25G
Tetrahexylammonium hydrogen sulphate ≥98.0% (by total nitrogen and titration analysis)
Tetrahexylammonium hydrogen sulphate
Formel: C₂₄H₅₃NO₄S
MW: 451,76 g/mol
Smeltepunkt: 98°C
MDL nummer: MFCD00037675
CAS-nummer: 32503-34-7
EINECS: 251-069-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b052f9f468759e53fde7e68ddc8789476b607121&vid=b052f9f468759e53fde7e68ddc8789476b607121&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR