Tyrphostin AG528 ≥97.0% (by HPLC)
Leverandør: TCI
|
Warning
|
Synonymer:
(E)-3-(3,4-Dihydroxyphenyl)-2-(indoline-1-carbonyl)acrylonitrile
T3504-25MG
T3504-100MG
TCIAT3504-25MGEA
2250
NOK
TCIAT3504-25MG
TCIAT3504-100MG
Tyrphostin AG528 ≥97.0% (by HPLC)
Tyrphostin AG528
Formel:
C₁₈H₁₄N₂O₃ MW: 306,32 g/mol Smeltepunkt: 215°C Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD01076474 CAS-nummer: 133550-49-9 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 8.60 to 9.60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...