Tris(trimethylsiloxy)silane ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxane, 1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]trisiloxane, Tris(trimethylsilyloxy)silane
T3520-5ML
T3520-25ML
TCIAT3520-5MLEA
340
NOK
TCIAT3520-5ML
TCIAT3520-25ML
Tris(trimethylsiloxy)silane ≥98.0% (by GC)
Tris(trimethylsiloxy)silane
Formel:
C₉H₂₈O₃Si₄ MW: 296,66 g/mol Kokepunkt: 185°C Flash punkt: 55°C |
MDL nummer:
MFCD00054865 CAS-nummer: 1873-89-8 EINECS: 217-497-7 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...