Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Tris(1,10-phenanthroline)cobalt bis(hexafluorophosphate)

T3553-1G T3553-5G
TCIAT3553-1GEA 1300 NOK
TCIAT3553-1G TCIAT3553-5G
Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)
Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate)
Formel: C₃₆H₂₄CoF₁₂N₆P₂
MW: 889,49 g/mol
MDL nummer: MFCD30749310
CAS-nummer: 31876-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb4da42904ef252edcf5821e47840d8b2d93fd39&vid=fb4da42904ef252edcf5821e47840d8b2d93fd39&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 98.0 %
Solubility in Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR