[(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥97.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: (4-(Trifluoromethyl)phenyl)(2,4,6-trimethoxyphenyl)iodonium 4-methylbenzenesulphonate, [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate

T3622-200MG T3622-1G
TCIAT3622-200MGEA 1050 NOK
TCIAT3622-200MG TCIAT3622-1G
[(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥97.0% (by HPLC)
[(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate
Formel: C₂₃H₂₂F₃IO₆S
MW: 610,38 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD30725767
CAS-nummer: 1868173-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7bb68aea5ce05316d965cd73abe86f18cb541b0&vid=c7bb68aea5ce05316d965cd73abe86f18cb541b0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 97.0 % (total of Ion-Pair)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR