1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: Isocyanuric acid tris(3,5-di-tert-butyl-4-hydroxybenzyl) ester, Tris(3,5-di-tert-butyl-4-hydroxybenzyl) Isocyanurate

T3653-100G T3653-500G
TCIAT3653-100GEA 1050 NOK
TCIAT3653-100G TCIAT3653-500G
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione ≥98.0% (by HPLC, total nitrogen)
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione
Formel: C₄₈H₆₉N₃O₆
MW: 784,10 g/mol
MDL nummer: MFCD00134700
CAS-nummer: 27676-62-6
EINECS: 248-597-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65ff6441bcc958b28d5e8d5fc5801fb70b3a1f98&vid=65ff6441bcc958b28d5e8d5fc5801fb70b3a1f98&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 220.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR