2-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 2-Methoxybenzeneboronic acid pinacol ester, 2-Methoxyphenylboronic acid, pinacol ester, 2-(2-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3708-25G T3708-5G
TCIAT3708-25GEA 2900 NOK
TCIAT3708-25G TCIAT3708-5G
2-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Methoxyphenylboronic acid pinacol ester
Formel: C₁₃H₁₉BO₃
MW: 234,10 g/mol
MDL nummer: MFCD05863916
CAS-nummer: 190788-60-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR