(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer:
(E,E,E,E)-4,4',4'',4'''-[Benzene-1,2,4,5-tetrayltetrakis(ethene-2,1-diyl)]tetrakis[N,N-bis(4-methoxyphenyl)aniline], TOP-HTM-α2
T3722-1G
T3722-25G
T3722-5G
TCIAT3722-1GEA
960
NOK
TCIAT3722-1G
TCIAT3722-25G
TCIAT3722-5G
(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
(E,E,E,E)-1,2,4,5-Tetrakis[4-[bis(4-methoxyphenyl)amino]styryl]benzene
Formel:
C₉₄H₈₂N₄O₈ MW: 1395,71 g/mol |
CAS-nummer:
2411528-61-3 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...