4-(Carboxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-[(4-Carboxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid, 4-(Carboxymethyl)phenylboronic acid Pinacol ester

T3754-200MG T3754-1G
TCIAT3754-200MGEA 420 NOK
TCIAT3754-200MG TCIAT3754-1G
4-(Carboxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Carboxymethyl)phenylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₄
MW: 262,1093 g/mol
MDL nummer: MFCD04038755
CAS-nummer: 797755-07-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR